missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CLCNKB Partial ORF (NP_000076.1, 516 a.a. - 615 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00001188-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16196404

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Chloride channel Kb (CLCNKB) is a member of the CLC family of voltage-gated chloride channels, which comprises at least 9 mammalian chloride channels. Each is believed to have 12 transmembrane domains and intracellular N and C termini. Mutations in CLCNKB result in the autosomal recessive Type III Bartter Syndrome. CLCNKA and CLCNKB are closely related (94% sequence identity), tightly linked (separated by 11 kb of genomic sequence) and are both expressed in mammalian kidney. [provided by RefSeq]

Sequence: QPSFYDGTVIVKKLPYLPRILGRNIGSHRVRVEHFMNHSITTLAKDTPLEEVVKVVTSTDVAEYPLVESTESQILVGIVRRAQLVQALKAEPPSWAPGHQ
Especificaciones

Especificaciones

NP_000076.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QPSFYDGTVIVKKLPYLPRILGRNIGSHRVRVEHFMNHSITTLAKDTPLEEVVKVVTSTDVAEYPLVESTESQILVGIVRRAQLVQALKAEPPSWAPGHQ
RUO
CLCNKB
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
1188
CLCNKB (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CLCKB/ClC-K2/MGC24087/hClC-Kb
CLCNKB
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales