Learn More
Abnova™ Human CLCN5 Partial ORF (NP_000075, 566 a.a. - 665 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00001184-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the ClC family of chloride ion channels and ion transporters. Mutations in this gene have been found in Dent disease and renal tubular disorders complicated by nephrolithiasis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: GYPFLEAKEEFAHKTLAMDVMKPRRNDPLLTVLTQDSMTVEDVETIISETTYSGFPVVVSRESQRLVGFVLRRDLIISIENARKKQDGVVSTSIIYFTEHEspecificaciones
NP_000075 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GYPFLEAKEEFAHKTLAMDVMKPRRNDPLLTVLTQDSMTVEDVETIISETTYSGFPVVVSRESQRLVGFVLRRDLIISIENARKKQDGVVSTSIIYFTEH | |
RUO | |
CLCN5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1184 | |
CLCN5 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLC5/CLCK2/DENTS/NPHL1/NPHL2/XLRH/XRN/hCIC-K2/hClC-K2 | |
CLCN5 | |
Recombinant | |
wheat germ expression system |