Learn More
Abnova™ Human CLCA3 Full-length ORF (AAI46528.1, 1 a.a. - 262 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009629-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is a transcribed pseudogene belonging to the calcium sensitive chloride conductance protein family. To date, all members of this gene family map to the same site on chromosome 1p31-p22 and share high degrees of homology in size, sequence and predicted structure, but differ significantly in their tissue distributions. This gene contains several nonsense codons compared to other family members that render the transcript a candidate for nonsense-mediated mRNA decay (NMD). Therefore, this gene is unlikely to be protein-coding. [provided by RefSeq]
Sequence: MVFSLKVILFLSLLLSPVLKSSLVTLNNNGYDGIVIAINPSVPEDEKLIQNIKEMVTEASTYLFHATKQRAYFRNVSILIPMTYKSKSEYLIPKQESYDQADVIVADLYLKYGDDPYTLQYGQCGDKGQYIHFTPNFLLTNNLATYGPRGRVFVHEWAHLRWGVFDEYNVDQPFYISRRNTTEATRCSTRITVYMVLNECKGASCIARPFRRDSKTGLYEAKCTFIPKRSQTAKESIVFMQNLDSVTEFCTEKTHNKEAPNLEspecificaciones
AAI46528.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.77kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLCA3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9629 | |
CLCA3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVFSLKVILFLSLLLSPVLKSSLVTLNNNGYDGIVIAINPSVPEDEKLIQNIKEMVTEASTYLFHATKQRAYFRNVSILIPMTYKSKSEYLIPKQESYDQADVIVADLYLKYGDDPYTLQYGQCGDKGQYIHFTPNFLLTNNLATYGPRGRVFVHEWAHLRWGVFDEYNVDQPFYISRRNTTEATRCSTRITVYMVLNECKGASCIARPFRRDSKTGLYEAKCTFIPKRSQTAKESIVFMQNLDSVTEFCTEKTHNKEAPNL | |
RUO | |
CLCA3 | |
Recombinant | |
wheat germ expression system |