Learn More
Abnova™ Human CHODL Partial ORF (NP_079220, 23 a.a. - 94 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00140578-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a type I membrane protein with a carbohydrate recognition domain characteristic of C-type lectins in its extracellular portion. In other proteins, this domain is involved in endocytosis of glycoproteins and exogenous sugar-bearing pathogens. This protein localizes predominantly to the perinuclear region. [provided by RefSeq]
Sequence: RVVSGQKVCFADFKHPCYKMAYFHELSSRVSFQEARLACESEGGVLLSLENEAEQKLIESMLQNLTKPGTGIEspecificaciones
NP_079220 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.66kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RVVSGQKVCFADFKHPCYKMAYFHELSSRVSFQEARLACESEGGVLLSLENEAEQKLIESMLQNLTKPGTGI | |
RUO | |
CHODL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
140578 | |
CHODL (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C21orf68/FLJ12627/MT75/PRED12 | |
CHODL | |
Recombinant | |
wheat germ expression system |