Learn More
Abnova™ Human CENPP Full-length ORF (NP_001012267.1, 1 a.a. - 288 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00401541-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
CENPP is a subunit of a CENPH (MIM 605607)-CENPI (MIM 300065)-associated centromeric complex that targets CENPA (MIM 117139) to centromeres and is required for proper kinetochore function and mitotic progression (Okada et al., 2006 [PubMed 16622420]).[supplied by OMIM]
Sequence: MDAELAEVRALQAEIAALRRACEDPPAPWEEKSRVQKSFQAIHQFNLEGWKSSKDLKNQLGHLESELSFLSTLTGINIRNHSKQTEDLTSTEMTEKSIRKVLQRHRLSGNCHMVTFQLEFQILEIQNKERLSSAVTDLNIIMEPTECSELSEFVSRAEERKDLFMFFRSLHFFVEWFEYRKRTFKHLKEKYPDAVYLSEGPSSCSMGIRSASRPGFELVIVWRIQIDEDGKVFPKLDLLTKVPQRALELDKNRAIETAPLSFRTLVGLLGIEAALESLIKSLCAEENNEspecificaciones
NP_001012267.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CENP-P/FLJ33928 | |
CENPP | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
401541 | |
CENPP (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDAELAEVRALQAEIAALRRACEDPPAPWEEKSRVQKSFQAIHQFNLEGWKSSKDLKNQLGHLESELSFLSTLTGINIRNHSKQTEDLTSTEMTEKSIRKVLQRHRLSGNCHMVTFQLEFQILEIQNKERLSSAVTDLNIIMEPTECSELSEFVSRAEERKDLFMFFRSLHFFVEWFEYRKRTFKHLKEKYPDAVYLSEGPSSCSMGIRSASRPGFELVIVWRIQIDEDGKVFPKLDLLTKVPQRALELDKNRAIETAPLSFRTLVGLLGIEAALESLIKSLCAEENN | |
RUO | |
CENPP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |