Learn More
Abnova™ Human CDS1 Partial ORF (NP_001254, 1 a.a. - 98 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00001040-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Breakdown products of phosphoinositides are ubiquitous second messengers that function downstream of many G protein-coupled receptors and tyrosine kinases regulating cell growth, calcium metabolism, and protein kinase C activity. This gene encodes an enzyme which regulates the amount of phosphatidylinositol available for signaling by catalyzing the conversion of phosphatidic acid to CDP-diacylglycerol. This enzyme is an integral membrane protein localized to two subcellular domains, the matrix side of the inner mitochondrial membrane where it is thought to be involved in the synthesis of phosphatidylglycerol and cardiolipin and the cytoplasmic side of the endoplasmic reticulum where it functions in phosphatidylinositol biosynthesis. Two genes encoding this enzyme have been identified in humans, one mapping to human chromosome 4q21 and a second to 20p13. [provided by RefSeq]
Sequence: MLELRHRGSCPGPREAVSPPHREGEAAGGDHETESTSDKETDIDDRYGDLDSRTDSDIPEIPPSSDRTPEILKKALSGLSSRWKNWWIRGILTLTMISEspecificaciones
NP_001254 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLELRHRGSCPGPREAVSPPHREGEAAGGDHETESTSDKETDIDDRYGDLDSRTDSDIPEIPPSSDRTPEILKKALSGLSSRWKNWWIRGILTLTMIS | |
RUO | |
CDS1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1040 | |
CDS1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDS | |
CDS1 | |
Recombinant | |
wheat germ expression system |