missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CDH8 Full-length ORF (NP_001787.2, 1 a.a. - 799 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00001006-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16192981

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a type II classical cadherin from the cadherin superfamily, integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Mature cadherin proteins are composed of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small, highly conserved C-terminal cytoplasmic domain. The extracellular domain consists of 5 subdomains, each containing a cadherin motif, and appears to determine the specificity of the protein's homophilic cell adhesion activity. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. This particular cadherin is expressed in brain and is putatively involved in synaptic adhesion, axon outgrowth and guidance. [provided by RefSeq]

Sequence: MPERLAEMLLDLWTPLIILWITLPPCIYMAPMNQSQVLMSGSPLELNSLGEEQRILNRSKRGWVWNQMFVLEEFSGPEPILVGRLHTDLDPGSKKIKYILSGDGAGTIFQINDVTGDIHAIKRLDREEKAEYTLTAQAVDWETSKPLEPPSEFIIKVQDINDNAPEFLNGPYHATVPEMSILGTSVTNVTATDADDPVYGNSAKLVYSILEGQPYFSIEPETAIIKTALPNMDREAKEEYLVVIQAKDMGGHSGGLSGTTTLTVTLTDVNDNPPKFAQSLYHFSVPEDVVLGTAIGRVKANDQDIGENAQSSYDIIDGDGTALFEITSDAQAQDGIIRLRKPLDFETKKSYTLKVEAANVHIDPRFSGRGPFKDTATVKIVVEDADEPPVFSSPTYLLEVHENAALNSVIGQVTARDPDITSSPIRFSIDRHTDLERQFNINADDGKITLATPLDRELSVWHNITIIATEIRNHSQISRVPVAIKVLDVNDNAPEFASEYEAFLCENGKPGQVIQTVSAMDKDDPKNGHYFLYSLLPEMVNNPNFTIKKNEDNSLSILAKHNGFNRQKQEVYLLPIIISDSGNPPLSSTSTLTIRVCGCSNDGVVQSCNVEAYVLPIGLSMGALIAILACIILLLVIVVLFVTLRRHKNEPLIIKDDEDVRENIIRYDDEGGGEEDTEAFDIATLQNPDGINGFLPRKDIKPDLQFMPRQGLAPVPNGVDVDEFINVRLHEADNDPTAPPYDSIQIYGYEGRGSVAGSLSSLESTTSDSDQNFDYLSDWGPRFKRLGELYSVGESDKET
Especificaciones

Especificaciones

NP_001787.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
114.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Nbla04261
CDH8
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
1006
CDH8 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPERLAEMLLDLWTPLIILWITLPPCIYMAPMNQSQVLMSGSPLELNSLGEEQRILNRSKRGWVWNQMFVLEEFSGPEPILVGRLHTDLDPGSKKIKYILSGDGAGTIFQINDVTGDIHAIKRLDREEKAEYTLTAQAVDWETSKPLEPPSEFIIKVQDINDNAPEFLNGPYHATVPEMSILGTSVTNVTATDADDPVYGNSAKLVYSILEGQPYFSIEPETAIIKTALPNMDREAKEEYLVVIQAKDMGGHSGGLSGTTTLTVTLTDVNDNPPKFAQSLYHFSVPEDVVLGTAIGRVKANDQDIGENAQSSYDIIDGDGTALFEITSDAQAQDGIIRLRKPLDFETKKSYTLKVEAANVHIDPRFSGRGPFKDTATVKIVVEDADEPPVFSSPTYLLEVHENAALNSVIGQVTARDPDITSSPIRFSIDRHTDLERQFNINADDGKITLATPLDRELSVWHNITIIATEIRNHSQISRVPVAIKVLDVNDNAPEFASEYEAFLCENGKPGQVIQTVSAMDKDDPKNGHYFLYSLLPEMVNNPNFTIKKNEDNSLSILAKHNGFNRQKQEVYLLPIIISDSGNPPLSSTSTLTIRVCGCSNDGVVQSCNVEAYVLPIGLSMGALIAILACIILLLVIVVLFVTLRRHKNEPLIIKDDEDVRENIIRYDDEGGGEEDTEAFDIATLQNPDGINGFLPRKDIKPDLQFMPRQGLAPVPNGVDVDEFINVRLHEADNDPTAPPYDSIQIYGYEGRGSVAGSLSSLESTTSDSDQNFDYLSDWGPRFKRLGELYSVGESDKET
RUO
CDH8
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales