missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CD155 (PVR) Fc Chimera, Recombinant Protein
HEK293 cells Recombinant Protein
Marca: Invitrogen™ A42501
Detalles adicionales : Peso : 0.01820kg
Descripción
The protein encoded by this gene is a transmembrane glycoprotein belonging to the immunoglobulin superfamily. The external domain mediates cell attachment to the extracellular matrix molecule vitronectin, while its intracellular domain interacts with the dynein light chain Tctex-1/DYNLT1. The gene is specific to the primate lineage, and serves as a cellular receptor for poliovirus in the first step of poliovirus replication. Multiple transcript variants encoding different isoforms have been found for this gene.Especificaciones
P15151 | |
PBS with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | |
Human CD155 (PVR) Fc Chimera | |
HEK293 cells | |
RUO | |
3830421F03Rik; CD155; D7Ertd458e; FLJ25946; HVED; mE4; NECL5; NECL-5; nectin-like 5; Nectin-like protein 5; poliovirus receptor; PVR; PVS; sCD112; soluble CD112; Taa1; TAGE4; tumor-associated antigen 1; tumor-associated glycoprotein pE4 | |
Human | |
Recombinant | |
≥ 90% |
Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot | |
5817 | |
5 x 50 μg | |
-20°C | |
<1 EU/μg | |
PVR | |
Unconjugated | |
Lyophilized |