missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CCDC102B Full-length ORF (NP_079057.1, 1 a.a. - 297 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079839-P01.25ug

Additional Details : Peso : 0.00010kg

 View more versions of this product

Product Code. 16104543

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 27-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Description

Description

Sequence: MKPNLDGVDLFNNGGSGNGETKTGLRLKAINLPLENEVTEISALQVHLDEFQKILWKEREMRTALEKEIERLESALSLWKWKYEELKESKPKNVKEFDILLGQHNDEMQELSGNIKEESKSQNSKDRVICELRAELERLQAENTSEWDKREILEREKQGLERENRRLKIQVKEMEELLDKKNRLSANSQSPDFKMSQIDLQEKNQELLNLQHAYYKLNRQYQANIAELTHANNRVDQNEAEVKKLRLRVEELKQGLNQKEDELDDSLNQIRKLQRSLDEEKERNENLETELRHLQNW
Specifications

Specifications

NP_079057.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
61.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACY1L/AN/C18orf14/DKFZp434K1426/DKFZp686I08254/FLJ23594/HsT1731/MGC161726/MGC161728
CCDC102B
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
79839
CCDC102B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKPNLDGVDLFNNGGSGNGETKTGLRLKAINLPLENEVTEISALQVHLDEFQKILWKEREMRTALEKEIERLESALSLWKWKYEELKESKPKNVKEFDILLGQHNDEMQELSGNIKEESKSQNSKDRVICELRAELERLQAENTSEWDKREILEREKQGLERENRRLKIQVKEMEELLDKKNRLSANSQSPDFKMSQIDLQEKNQELLNLQHAYYKLNRQYQANIAELTHANNRVDQNEAEVKKLRLRVEELKQGLNQKEDELDDSLNQIRKLQRSLDEEKERNENLETELRHLQNW
RUO
CCDC102B
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions