Learn More
Abnova™ Human CALB2 Full-length ORF (NP_009018.1, 1 a.a. - 179 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00000794-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes an intracellular calcium-binding protein belonging to the troponin C superfamily. Members of this protein family have six EF-hand domains which bind calcium. Three alternatively spliced transcript variants that encode different proteins have been described. [provided by RefSeq]
Sequence: MAGPQQQPPYLHLAELTASQFLEIWKHFDADGNGYIEGKELENFFQELEKARKGSGMMSKSDNFGEKMKEFMQKYDKNSDGKIEMAELAQILPTEENFLLCFRQHVGSSAEFMEAWRKYDTDRSGYIEANELKGFLSDLLKKANRPYDEPKLQEYTQTILRMFDLNGDGKLGLSEMSRAEspecificaciones
NP_009018.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
47.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAL2 | |
CALB2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
794 | |
CALB2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAGPQQQPPYLHLAELTASQFLEIWKHFDADGNGYIEGKELENFFQELEKARKGSGMMSKSDNFGEKMKEFMQKYDKNSDGKIEMAELAQILPTEENFLLCFRQHVGSSAEFMEAWRKYDTDRSGYIEANELKGFLSDLLKKANRPYDEPKLQEYTQTILRMFDLNGDGKLGLSEMSRA | |
RUO | |
CALB2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |