missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CACNB4 Full-length ORF (NP_000717.2, 1 a.a. - 520 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00000785-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16102521

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a member of the beta subunit family of voltage-dependent calcium channel complex proteins. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Various versions of each of these subunits exist, either expressed from similar genes or the result of alternative splicing. The protein encoded by this locus plays an important role in calcium channel function by modulating G protein inhibition, increasing peak calcium current, controlling the alpha-1 subunit membrane targeting and shifting the voltage dependence of activation and inactivation. Certain mutations in this gene have been associated with idiopathic generalized epilepsy (IGE) and juvenile myoclonic epilepsy (JME). Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MSSSSYAKNGTADGPHSPTSQVARGTTTRRSRLKRSDGSTTSTSFILRQGSADSYTSRPSDSDVSLEEDREAIRQEREQQAAIQLERAKSKPVAFAVKTNVSYCGALDEDVPVPSTAISFDAKDFLHIKEKYNNDWWIGRLVKEGCEIGFIPSPLRLENIRIQQEQKRGRFHGGKSSGNSSSSLGEMVSGTFRATPTSTAKQKQKVTEHIPPYDVVPSMRPVVLVGPSLKGYEVTDMMQKALFDFLKHRFDGRISITRVTADISLAKRSVLNNPSKRAIIERSNTRSSLAEVQSEIERIFELARSLQLVVLDADTINHPAQLIKTSLAPIIVHVKVSSPKVLQRLIKSRGKSQSKHLNVQLVAADKLAQCPPEMFDVILDENQLEDACEHLGEYLEAYWRATHTTSSTPMTPLLGRNLGSTALSPYPTAISGLQSQRMRHSNHSTENSPIERRSLMTSDENYHNERARKSRNRLSSSSQHSRDHYPLVEEDYPDSYQDTYKPHRNRGSPGGYSHDSRHRL
Especificaciones

Especificaciones

NP_000717.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
84.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAB4/CACNLB4/EA5/EJM
CACNB4
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
785
CACNB4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSSSSYAKNGTADGPHSPTSQVARGTTTRRSRLKRSDGSTTSTSFILRQGSADSYTSRPSDSDVSLEEDREAIRQEREQQAAIQLERAKSKPVAFAVKTNVSYCGALDEDVPVPSTAISFDAKDFLHIKEKYNNDWWIGRLVKEGCEIGFIPSPLRLENIRIQQEQKRGRFHGGKSSGNSSSSLGEMVSGTFRATPTSTAKQKQKVTEHIPPYDVVPSMRPVVLVGPSLKGYEVTDMMQKALFDFLKHRFDGRISITRVTADISLAKRSVLNNPSKRAIIERSNTRSSLAEVQSEIERIFELARSLQLVVLDADTINHPAQLIKTSLAPIIVHVKVSSPKVLQRLIKSRGKSQSKHLNVQLVAADKLAQCPPEMFDVILDENQLEDACEHLGEYLEAYWRATHTTSSTPMTPLLGRNLGSTALSPYPTAISGLQSQRMRHSNHSTENSPIERRSLMTSDENYHNERARKSRNRLSSSSQHSRDHYPLVEEDYPDSYQDTYKPHRNRGSPGGYSHDSRHRL
RUO
CACNB4
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales