missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human C14orf131 Full-length ORF (NP_060805.2, 1 a.a. - 428 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055778-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MKVEKDHLAKPFFPAIYKEFEELHKLVKKMCQDYLSSSGLCSQETLEINNDKVAESLGITEFLRKKEIHPDNLGPKHLSRDMDGEQLEGASSEKREREAAEEGLASVKRPRREALSNDTTESLAANSRGREKPRPLHALAAGEGFSPPVNVTVSPRSEESHTTTVSGGNGSVFQAGPQLQALANLEARRGSIGAALSSRDVSGLPVYAQSGEPRRLTQAQVAAFPGENALEHSSDQDTWDSLRSPGFCSPLSSGGGAESLPPGGPGHAEAGHLGKVCDFHLNHQQPSPTSVLPTEVAAPPLEKILSVDSVAVDCAYRTVPKPGPQPGPHGSLLTEGCLRSLSGDLNRFPCGMEVHSGQRELESVVAVGEAMAFEISNGSHELLSQGQKQIFIQTSDGLILSPPGTIVSQEEDIVTVTDAEGRACGWAREspecificaciones
NP_060805.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
72kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C14orf131/FLJ11132 | |
ZNF839 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55778 | |
C14orf131 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKVEKDHLAKPFFPAIYKEFEELHKLVKKMCQDYLSSSGLCSQETLEINNDKVAESLGITEFLRKKEIHPDNLGPKHLSRDMDGEQLEGASSEKREREAAEEGLASVKRPRREALSNDTTESLAANSRGREKPRPLHALAAGEGFSPPVNVTVSPRSEESHTTTVSGGNGSVFQAGPQLQALANLEARRGSIGAALSSRDVSGLPVYAQSGEPRRLTQAQVAAFPGENALEHSSDQDTWDSLRSPGFCSPLSSGGGAESLPPGGPGHAEAGHLGKVCDFHLNHQQPSPTSVLPTEVAAPPLEKILSVDSVAVDCAYRTVPKPGPQPGPHGSLLTEGCLRSLSGDLNRFPCGMEVHSGQRELESVVAVGEAMAFEISNGSHELLSQGQKQIFIQTSDGLILSPPGTIVSQEEDIVTVTDAEGRACGWAR | |
RUO | |
ZNF839 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |