Learn More
Abnova™ Human BRWD1 Full-length ORF (NP_001007247.1, 1 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00054014-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This protein contains 2 bromodomains and multiple WD repeats, and the function of this protein is not known. This gene is located within the Down syndrome region-2 on chromosome 21. Alternative splicing of this gene generates 3 transcript variants diverging at the 3' ends. [provided by RefSeq]
Sequence: MAEPSSARRPVPLIESELYFLIARYLSAGPCRRAAQVLVQELEQYQLLPKRLDWEGNEHNRSYEELVLSNKHVAPDHLLQICQRIGPMLDKEIPPSISRVTSLLGAGRQSLLRTAKGTLIEspecificaciones
NP_001007247.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
40kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
BRWD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
54014 | |
BRWD1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C21orf107/FLJ43918/N143/WDR9 | |
BRWD1 | |
Recombinant | |
wheat germ expression system |