missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BRS3 Partial ORF (NP_001718, 290 a.a. - 399 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00000680-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16192341

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Mammalian bombesin-like peptides (see MIM 137260) are widely distributed in the central nervous system as well as in the gastrointestinal tract, where they modulate smooth-muscle contraction, exocrine and endocrine processes, metabolism, and behavior. They bind to G protein-coupled receptors on the cell surface to elicit their effects. Bombesin-like peptide receptors include gastrin-releasing peptide receptor (MIM 305670), neuromedin B receptor (MIM 162341), and bombesin-like receptor-3 (BRS3) (Ohki-Hamazaki et al., 1997).[supplied by OMIM]

Sequence: LYLYHSFTSQTYVDPSAMHFIFTIFSRVLAFSNSCVNPFALYWLSKSFQKHFKAQLFCCKAERPEPPVADTSLTTLAVMGTVPGTGSIQMSEISVTSFTGCSVKQAEDRF
Especificaciones

Especificaciones

NP_001718
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LYLYHSFTSQTYVDPSAMHFIFTIFSRVLAFSNSCVNPFALYWLSKSFQKHFKAQLFCCKAERPEPPVADTSLTTLAVMGTVPGTGSIQMSEISVTSFTGCSVKQAEDRF
RUO
BRS3
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
680
BRS3 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BRS3
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales