Learn More
Abnova™ Human BRF2 Partial ORF (NP_060780, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055290-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes one of the multiple subunits of the RNA polymerase III transcription factor complex required for transcription of genes with promoter elements upstream of the initiation site. The product of this gene, a TFIIB-like factor, is directly recruited to the TATA-box of polymerase III small nuclear RNA gene promoters through its interaction with the TATA-binding protein. [provided by RefSeq]
Sequence: MPGRGRCPDCGSTELVEDSHYSQSQLVCSDCGCVVTEGVLTTTFSDEGNLREVTYSRSTGENEQVSRSQQRGLRRVRDLCRVLQLPPTFEDTAVAYYQQAEspecificaciones
NP_060780 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPGRGRCPDCGSTELVEDSHYSQSQLVCSDCGCVVTEGVLTTTFSDEGNLREVTYSRSTGENEQVSRSQQRGLRRVRDLCRVLQLPPTFEDTAVAYYQQA | |
RUO | |
BRF2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55290 | |
BRF2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BRFU/FLJ11052/TFIIIB50 | |
BRF2 | |
Recombinant | |
wheat germ expression system |