Learn More
Abnova™ Human BLMH Full-length ORF (NP_000377.1, 1 a.a. - 455 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00000642-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Bleomycin hydrolase (BMH) is a cytoplasmic cysteine peptidase that is highly conserved through evolution; however, the only known activity of the enzyme is metabolic inactivation of the glycopeptide bleomycin (BLM), an essential component of combination chemotherapy regimens for cancer. The protein contains the signature active site residues of the cysteine protease papain superfamily. [provided by RefSeq]
Sequence: MSSSGLNSEKVAALIQKLNSDPQFVLAQNVGTTHDLLDICLKRATVQRAQHVFQHAVPQEGKPITNQKSSGRCWIFSCLNVMRLPFMKKLNIEEFEFSQSYLFFWDKVERCYFFLSAFVDTAQRKEPEDGRLVQFLLMNPANDGGQWDMLVNIVEKYGVIPKKCFPESYTTEATRRMNDILNHKMREFCIRLRNLVHSGATKGEISATQDVMMEEIFRVVCICLGNPPETFTWEYRDKDKNYQKIGPITPLEFYREHVKPLFNMEDKICLVNDPRPQHKYNKLYTVEYLSNMVGGRKTLYNNQPIDFLKKMVAASIKDGEAVWFGCDVGKHFNSKLGLSDMNLYDHELVFGVSLKNMNKAERLTFGESLMTHAMTFTAVSEKDDQDGAFTKWRVENSWGEDHGHKGYLCMTDEWFSEYVYEVVVDRKHVPEEVLAVLEQEPIILPAWDPMGALAEEspecificaciones
NP_000377.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
79kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
BLMH | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
642 | |
BLMH (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BH/BMH | |
BLMH | |
Recombinant | |
wheat germ expression system |