missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human BHLHB8 Partial ORF (NP_803238.1, 75 a.a. - 140 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00168620-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: QRRLESNERERQRMHKLNNAFQALREVIPHVRADKKLSKIETLTLAKNYIKSLTATILTMSSSRLPEspecificaciones
NP_803238.1 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QRRLESNERERQRMHKLNNAFQALREVIPHVRADKKLSKIETLTLAKNYIKSLTATILTMSSSRLP | |
RUO | |
BHLHA15 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
168620 | |
BHLHB8 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BHLHB8|MIST1 | |
BHLHA15 | |
Yes |