missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BCS1L Partial ORF (NP_004319, 320 a.a. - 418 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00000617-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16195184

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a homolog of the S. cerevisiae bcs1 protein which is involved in the assembly of complex III of the mitochondrial respiratory chain. The encoded protein does not contain a mitochondrial targeting sequence but experimental studies confirm that it is imported into mitochondria. Mutations in this gene are associated with mitochondrial complex III deficiency and the GRACILE syndrome. Two alternatively spliced transcripts encoding the same protein have been described. [provided by RefSeq]

Sequence: ASTEARIVFMTTNHVDRLDPALIRPGRVDLKEYVGYCSHWQLTQMFQRFYPGQAPSLAENFAEHVLRATNQISPAQVQGYFMLYKNDPVGAIHNAESLR
Especificaciones

Especificaciones

NP_004319
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ASTEARIVFMTTNHVDRLDPALIRPGRVDLKEYVGYCSHWQLTQMFQRFYPGQAPSLAENFAEHVLRATNQISPAQVQGYFMLYKNDPVGAIHNAESLR
RUO
BCS1L
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
617
BCS1L (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BCS/BCS1/BJS/FLNMS/GRACILE/Hs.6719/PTD/h-BCS
BCS1L
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales