Learn More
Abnova™ Human BCL7A Full-length ORF (NP_001019979.1, 1 a.a. - 210 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00000605-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is directly involved, with Myc and IgH, in a three-way gene translocation in a Burkitt lymphoma cell line. As a result of the gene translocation, the N-terminal region of the gene product is disrupted, which is thought to be related to the pathogenesis of a subset of high-grade B cell non-Hodgkin lymphoma. The N-terminal segment involved in the translocation includes the region that shares a strong sequence similarity with those of BCL7B and BCL7C. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MSGRSVRAETRSRAKDDIKRVMAAIEKVRKWEKKWVTVGDTSLRIYKWVPVTEPKVDDKNKNKKKGKDEKCGSEVTTPENSSSPGMMDMHDDNSNQSSIADASPIKQENSSNSSPAPEPNSAVPSDGTEAKVDEAQADGKEHPGAEDASDEQNSQSSMEHSMNSSEKVDRQPSGDSGLAAETSAISQDLEGVPPSKKMKLEASQQNSEEMEspecificaciones
NP_001019979.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
49.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
BCL7A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
605 | |
BCL7A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BCL7 | |
BCL7A | |
Recombinant | |
wheat germ expression system |