missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BCL2L11 Partial ORF (NP_619527, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00010018-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16191656

  • 310.00€ / 10 microgramos


Fecha estimada de envío
15-02-2021
Añadir a la cesta

Descripción

Descripción

Sequence: MAKQPSDVSSECDREGRQLQPAERPPQLRPGAPTSLQTEPQGNPEGNHGGEGDSCPHGSPQGPLAPPASPGPFATRSPLFIFMRRSSLLSRSSSGYFSFD
Especificaciones

Especificaciones

NP_619527
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MAKQPSDVSSECDREGRQLQPAERPPQLRPGAPTSLQTEPQGNPEGNHGGEGDSCPHGSPQGPLAPPASPGPFATRSPLFIFMRRSSLLSRSSSGYFSFD
RUO
BCL2L11
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
10018
BCL2L11 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BAM|BIM|BIM-alpha6|BIM-beta6|BIM-beta7|BOD|BimEL|BimL
BCL2L11
Yes
Videos
SDS
Documentos