Learn More
Abnova™ Human BCAS1 Partial ORF (NP_003648, 1 a.a. - 80 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008537-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene resides in a region at 20q13 which is amplified in a variety of tumor types and associated with more aggressive tumor phenotypes. Among the genes identified from this region, it was found to be highly expressed in three amplified breast cancer cell lines and in one breast tumor without amplification at 20q13.2. However, this gene is not in the common region of maximal amplification and its expression was not detected in the breast cancer cell line MCF7, in which this region is highly amplified. Although not consistently expressed, this gene is a candidate oncogene. [provided by RefSeq]
Sequence: MGNQMSVPQRVEDQENEPEAETYQDNASALNGVPVVVSTHTVQHLEEVDLGISVKTDNVATSSPETTEISAVADANGKNLEspecificaciones
NP_003648 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.54kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGNQMSVPQRVEDQENEPEAETYQDNASALNGVPVVVSTHTVQHLEEVDLGISVKTDNVATSSPETTEISAVADANGKNL | |
RUO | |
BCAS1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8537 | |
BCAS1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AIBC1/NABC1 | |
BCAS1 | |
Recombinant | |
wheat germ expression system |