missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BCAR3 Partial ORF (AAH39895, 266 a.a. - 373 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00008412-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16108515

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Breast tumors are initially dependent on estrogens for growth and progression and can be inhibited by anti-estrogens such as tamoxifen. However, breast cancers progress to become anti-estrogen resistant. Breast cancer anti-estrogen resistance gene 3 was identified in the search for genes involved in the development of estrogen resistance. The gene encodes a component of intracellular signal transduction that causes estrogen-independent proliferation in human breast cancer cells. The protein contains a putative src homology 2 (SH2) domain, a hall mark of cellular tyrosine kinase signaling molecules, and is partly homologous to the cell division cycle protein CDC48. [provided by RefSeq]

Sequence: YGTSPGQAREGSLTKGRPDVAKRLSLTMGGVQAREQNLPRGNLLRNKEKSGSQPACLDHMQDRRALSLKAHQSESYLPIGCKLPPQSSGVDTSPCPNSPVFRTGSEPA
Especificaciones

Especificaciones

AAH39895
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.51kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
YGTSPGQAREGSLTKGRPDVAKRLSLTMGGVQAREQNLPRGNLLRNKEKSGSQPACLDHMQDRRALSLKAHQSESYLPIGCKLPPQSSGVDTSPCPNSPVFRTGSEPA
RUO
BCAR3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8412
BCAR3 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0554/NSP2/SH2D3B
BCAR3
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales