Learn More
Abnova™ Human BAT5 Full-length ORF (NP_066983.1, 1 a.a. - 558 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00007920-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
A cluster of genes, BAT1-BAT5, has been localized in the vicinity of the genes for TNF alpha and TNF beta. These genes are all within the human major histocompatibility complex class III region. The protein encoded by this gene is thought to be involved in some aspects of immunity. [provided by RefSeq]
Sequence: MAKLLSCVLGPRLYKIYRERDSERAPASVPETPTAVTAPHSSSWDTYYQPRALEKHADSILALASVFWSISYYSSPFAFFYLYRKGYLSLSKVVPFSHYAGTLLLLLAGVACLRGIGRWTNPQYRQFITILEATHRNQSSENKRQLANYNFDFRSWPVDFHWEEPSSRKESRGGPSRRGVALLRPEPLHRGTADTLLNRVKKLPCQITSYLVAHTLGRRMLYPGSVYLLQKALMPVLLQGQARLVEECNGRRAKLLACDGNEIDTMFVDRRGTAEPQGQKLVICCEGNAGFYEVGCVSTPLEAGYSVLGWNHPGFAGSTGVPFPQNEANAMDVVVQFAIHRLGFQPQDIIIYAWSIGGFTATWAAMSYPDVSAMILDASFDDLVPLALKVMPDSWRGLVTRTVRQHLNLNNAEQLCRYQGPVLLIRRTKDEIITTTVPEDIMSNRGNDLLLKLLQHRYPRVMAEEGLRVVRQWLEASSQLEEASIYSRWEVEEDWCLSVLRSYQAEHGPDFPWSVGEDMSADGRRQLALFLARKHLHNFEATHCTPLPAQNFQMPWHLEspecificaciones
NP_066983.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
89.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
BAT5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7920 | |
BAT5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
D6S82E/NG26 | |
BAT5 | |
Recombinant | |
wheat germ expression system |