missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BAAT Partial ORF (NP_001692, 258 a.a. - 355 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00000570-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16195124

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene is a liver enzyme that catalyzes the transfer of C24 bile acids from the acyl-CoA thioester to either glycine or taurine, the second step in the formation of bile acid-amino acid conjugates. The bile acid conjugates then act as a detergent in the gastrointestinal tract, which enhances lipid and fat-soluble vitamin absorption. Defects in this gene are a cause of familial hypercholanemia (FHCA). Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]

Sequence: NGTNFPFGIPQVYHGQIHQPLPHSAQLISTNALGLLELYRTFETTQVGASQYLFPIEEAQGQFLFIVGEGDKTINSKAHAEQAIGQLKRHGKNNWTLL
Especificaciones

Especificaciones

NP_001692
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NGTNFPFGIPQVYHGQIHQPLPHSAQLISTNALGLLELYRTFETTQVGASQYLFPIEEAQGQFLFIVGEGDKTINSKAHAEQAIGQLKRHGKNNWTLL
RUO
BAAT
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
570
BAAT (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BACAT/BAT/FLJ20300/MGC104432
BAAT
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales