Learn More
Abnova™ Human AZI2 Full-length ORF (NP_071906.1, 1 a.a. - 392 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00064343-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
AZI2, or NAP1, contributes to the activation of NFKB (see MIM 164011)-dependent gene expression by activating IKK-related kinases, such as NAK (TBK1; MIM 604834) (Fujita et al., 2003 [PubMed 14560022]).[supplied by OMIM]
Sequence: MDALVEDDICILNHEKAHKRDTVTPVSIYSGDESVASHFALVTAYEDIKKRLKDSEKENSLLKKRIRFLEEKLIARFEEETSSVGREQVNKAYHAYREVCIDRDNLKSKLDKMNKDNSESLKVLNEQLQSKEVELLQLRTEVETQQVMRNLNPPSSNWEVEKLSCDLKIHGLEQELELMRKECSDLKIELQKAKQTDPYQEDNLKSRDLQKLSISSDNMQHAYWELKREMSNLHLVTQVQAELLRKLKTSTAIKKACAPVGCSEDLGRDSTKLHLMNFTATYTRHPPLLPNGKALCHTTSSPLPGDVKVLSEKAILQSWTDNERSIPNDGTCFQEHSSYGRNSLEDNSWVFPSPPKSSETAFGETKTKTLPLPNLPPLHYLDQHNQNCLYKNEspecificaciones
NP_071906.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
71.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
AZI2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
64343 | |
AZI2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AZ2/NAP1/TILP | |
AZI2 | |
Recombinant | |
wheat germ expression system |