missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ATP6V1E2 Full-length ORF (NP_542384.1, 1 a.a. - 226 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00090423-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16196493

  • 458.00€ / 25 microgramos


Fecha estimada de envío
19-10-2020
Añadir a la cesta

Descripción

Descripción

Sequence: MALSDVDVKKQIKHMMAFIEQEANEKAEEIDAKAEEEFNIEKGRLVQTQRLKIMEYYEKKEKQIEQQKKILMSTMRNQARLKVLRARNDLISDLLSEAKLRLSRIVEDPEVYQGLLDKLVLQGLLRLLEPVMIVRCRPQDLLLVEAAVQKAIPEYMTISQKHVEVQIDKEAYLAVNAAGGVEVYSGNQRIKVSNTLESRLDLSAKQKMPEIRMALFGANTNRKFFI
Especificaciones

Especificaciones

NP_542384.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
52.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
ATP6V1E2
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
90423
ATP6V1E2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATP6E1|ATP6EL2|ATP6V1EL2|MGC9341|VMA4
ATP6V1E2
Yes
Videos
Documentos