missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ATP6V1C1 Partial ORF (NP_001686, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00000528-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MTEFWLISAPGEKTCQQTWEKLHAATSKNNNLAVTSKFNIPDLKVGTLDVLVGLSDELAKLDAFVEGVVKKVAQYMADVLEDSKDKVQENLLANGVDLVTYITRFQWDMAEspecificaciones
NP_001686 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTEFWLISAPGEKTCQQTWEKLHAATSKNNNLAVTSKFNIPDLKVGTLDVLVGLSDELAKLDAFVEGVVKKVAQYMADVLEDSKDKVQENLLANGVDLVTYITRFQWDMA | |
RUO | |
ATP6V1C1 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
528 | |
ATP6V1C1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATP6C|ATP6D|FLJ20057|VATC|Vma5 | |
ATP6V1C1 | |
Yes |