missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ATP5G3 Full-length ORF (NP_001002258.1, 1 a.a. - 142 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00000518-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16102061

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel seems to have nine subunits (a, b, c, d, e, f, g, F6 and 8). This gene is one of three genes that encode subunit c of the proton channel. Each of the three genes have distinct mitochondrial import sequences but encode the identical mature protein. Alternatively spliced transcript variants encoding the same protein have been identified. [provided by RefSeq]

Sequence: MFACAKLACTPSLIRAGSRVAYRPISASVLSRPEASRTGEGSTVFNGAQNGVSQLIQREFQTSAISRDIDTAAKFIGAGAATVGVAGSGAGIGTVFGSLIIGYARNPSLKQQLFSYAILGFALSEAMGLFCLMVAFLILFAM
Especificaciones

Especificaciones

NP_001002258.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
41.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ATP5G3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
518
ATP5G3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC125738/P3
ATP5G3
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales