missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ATG4A Full-length ORF (NP_443168.2, 1 a.a. - 398 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00115201-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16197193

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 07-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Autophagy is the process by which endogenous proteins and damaged organelles are destroyed intracellularly. Autophagy is postulated to be essential for cell homeostasis and cell remodeling during differentiation, metamorphosis, non-apoptotic cell death, and aging. Reduced levels of autophagy have been described in some malignant tumors, and a role for autophagy in controlling the unregulated cell growth linked to cancer has been proposed. This gene encodes a member of the autophagin protein family. The encoded protein is also designated as a member of the C-54 family of cysteine proteases. Transcript variants that encode distinct isoforms have been identified. [provided by RefSeq]

Sequence: MESVLSKYEDQITIFTDYLEEYPDTDELVWILGKQHLLKTEKSKLLSDISARLWFTYRRKFSPIGGTGPSSDAGWGCMLRCGQMMLAQALICRHLGRDWSWEKQKEQPKEYQRILQCFLDRKDCCYSIHQMAQMGVGEGKSIGEWFGPNTVAQVLKKLALFDEWNSLAVYVSMDNTVVIEDIKKMCRVLPLSADTAGDRPPDSLTASNQSKGTSAYCSAWKPLLLIVPLRLGINQINPVYVDAFKECFKMPQSLGALGGKPNNAYYFIGFLGDELIFLDPHTTQTFVDTEENGTVNDQTFHCLQSPQRMNILNLDPSVALGFFCKEEKDFDNWCSLVQKEILKENLRMFELVQKHPSHWPPFVPPAKPEVTTTGAEFIDSTEQLEEFDLEEDFEILSV
Especificaciones

Especificaciones

NP_443168.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
71.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ATG4A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
115201
ATG4A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
APG4A/AUTL2
ATG4A
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales