missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ARL2BP Full-length ORF (AAH03087, 1 a.a. - 163 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00023568-P01S

 Ver más versiones de este producto

Código de producto. 16117482

  • 335.00€ / 10 microgramos

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Explore más ofertas especiales
Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

ADP-ribosylation factor (ARF)-like proteins (ARLs) comprise a functionally distinct group of the ARF family of RAS-related GTPases. The protein encoded by this gene binds to ARL2.GTP with high affinity but does not interact with ARL2.GDP, activated ARF, or RHO proteins. The lack of detectable membrane association of this protein or ARL2 upon activation of ARL2 is suggestive of actions distinct from those of the ARFs. This protein is considered to be the first ARL2-specific effector identified, due to its interaction with ARL2.GTP but lack of ARL2 GTPase-activating protein activity. [provided by RefSeq]

Sequence: MDALEGESFALSFSSASDAEFDAVVGYLEDIIMDDEFQLLQRNFMDKYYLEFEDTEENKLIYTPIFNEYISLVEKYIEEQLLQRIPEFNMAAFTTTLQHHKDEVAGDIFDMLLTFTDFLAFKEMFLDYRAEKEGRGLDLSSGLVVTSLCKSSSLPASQNNLRH
Especificaciones

Especificaciones

AAH03087
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
43.67kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
ARL2BP
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
23568
ARL2BP (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BART/BART1
ARL2BP
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales