missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human Apolipoprotein E / APOE Protein, His Tag

383.89€ - 2771.21€

Especificaciones

Peso molecular 36.1 kDa
Fuente Human Apolipoprotein E, His Tag (APE-H5246) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - His 317 (Accession # P02649-1).
Inmunógeno Lys 19 - His 317
Requisitos de almacenamiento -20°C
Alias de gen Apolipoprotein E,APOE,Apo-E
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
16477159
Ver documentos Detalles de la promoción
ACROBiosystems
APE-H5246-100UG
100 μg
383.89€
100 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 11-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16487159
Ver documentos Detalles de la promoción
ACROBiosystems
APE-H5246-1MG
1 mg
2771.21€
1 miligramo
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 11-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

APOE is an apolipoprotein, a protein associating with lipid particles, that mainly functions in lipoprotein-mediated lipid transport between organs via the plasma and interstitial fluids. APOE is a core component of plasma lipoproteins and is involved in their production, conversion and clearance. Apoliproteins are amphipathic molecules that interact both with lipids of the lipoprotein particle core and the aqueous environment of the plasma. As such, APOE associates with chylomicrons, chylomicron remnants, very low density lipoproteins (VLDL) and intermediate density lipoproteins (IDL) but shows a preferential binding to high-density lipoproteins (HDL). It also binds a wide range of cellular receptors including the LDL receptor/LDLR, the LDL receptor-related proteins LRP1, LRP2 and LRP8 and the very low-density lipoprotein receptor/VLDLR that mediate the cellular uptake of the APOE-containing lipoprotein particles.

This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 36.1 kDa. The protein migrates as 35-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones

Especificaciones

36.1 kDa
Lys 19 - His 317
Apolipoprotein E,APOE,Apo-E
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
Human Apolipoprotein E, His Tag (APE-H5246) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - His 317 (Accession # P02649-1).
-20°C
Apolipoprotein E
Human
PBS, 5 mM CHAPS, 2 mM DTT, pH 7.4
HEK293
Other Recombinant Proteins
Videos
SDS
Documentos

Documentos

Product Certifications