missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human APOL6 Full-length ORF (NP_085144.1, 1 a.a. - 343 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

en la oferta

Especificaciones

Nombre APOL6 (Human) Recombinant Protein (P01)
Recombinante Yes
Método de purificación Glutathione Sepharose 4 Fast Flow
Peso molecular 64.5kDa
Requisitos de almacenamiento Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Ver más especificaciones

Descripción

Descripción

This gene is a member of the apolipoprotein L gene family. The encoded protein is found in the cytoplasm, where it may affect the movement of lipids or allow the binding of lipids to organelles. [provided by RefSeq]

Sequence: MDNQAERESEAGVGLQRDEDDAPLCEDVELQDGDLSPEEKIFLREFPRLKEDLKGNIDKLRALADDIDKTHKKFTKANMVATSTAVISGVMSLLGLALAPATGGGSLLLSTAGQGLATAAGVTSIVSGTLERSKNKEAQARAEDILPTYDQEDREDEEEKADYVTAAGKIIYNLRNTLKYAKKNVRAFWKLRANPRLANATKRLLTTGQVSSRSRVQVQKAFAGTTLAMTKNARVLGGVMSAFSLGYDLATLSKEWKHLKEGARTKFAEELRAKALELERKLTELTQLYKSLQQKVRSRARGVGKDLTGTCETEAYWKELREHVWMWLWLCVCLCVCVYVQFT
Especificaciones

Especificaciones

APOL6 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
64.5kDa
Videos
SDS
Documentos

Documentos

Product Certifications