missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human APEX1 Partial ORF (NP_001632, 219 a.a. - 318 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00000328-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: DLRNPKGNKKNAGFTPQERQGFGELLQAVPLADSFRHLYPNTPYAYTFWTYMMNARSKNVGWRLDYFLLSHSLLPALCDSKIRSKALGSDHCPITLYLALEspecificaciones
NP_001632 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DLRNPKGNKKNAGFTPQERQGFGELLQAVPLADSFRHLYPNTPYAYTFWTYMMNARSKNVGWRLDYFLLSHSLLPALCDSKIRSKALGSDHCPITLYLAL | |
RUO | |
APEX1 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
328 | |
APEX1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APE|APE-1|APE1|APEN|APEX|APX|HAP1|REF-1|REF1 | |
APEX1 | |
Yes |