Learn More
Abnova™ Human AMBN Full-length ORF (NP_057603.1, 1 a.a. - 447 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00000258-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Description
Ameloblastin is thought to represent an unique ameloblast-specific gene product that may be important in enamel matrix formation and mineralization. The gene is located on chromosome 4 near other genes associated with mineralized tissues: osteopontin, bone sialoprotein, and bone morphogenetic protein 3. Based on its cytogenetic location, this gene is a candidate gene for one form of the disorder, dentinogenesis imperfecta, and/or the disorder, autosomal dominant amylogenesis imperfecta. [provided by RefSeq]
Sequence: MSASKIPLFKMKDLILILCLLEMSFAVPFFPQQSGTPGMASLSLETMRQLGSLQRLNTLSQYSRYGFGKSFNSLWMHGLLPPHSSLPWMRPREHETQQYEYSLPVHPPPLPSQPSLKPQQPGLKPFLQSAAATTNQATALKEALQPPIHLGHLPLQEGELPLVQQQVAPSDKPPKPELPGVDFADPQGPSLPGMDFPDPQGPSLPGLDFADPQGSTIFQIARLISHGPMPQNKQSPLYPGMLYVPFGANQLNAPARLGIMSSEEVAGGREDPMAYGAMFPGFGGMRPGFEGMPHNPAMGGDFTLEFDSPVAATKGPENEEGGAQGSPMPEANPDNLENPAFLTELEPAPHAGLLALPKDDIPGLPRSPSGKMKGLPSVTPAAADPLMTPELADVYRTYDADMTTSVDFQEEATMDTTMAPNSLQTSMPGNKAQEPEMMHDAWHFQEPSpecifications
NP_057603.1 | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
74.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
AMBN | |
Yes |
wheat germ expression system | |
Liquid | |
258 | |
AMBN (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AMBN | |
Wheat Germ (in vitro) | |
GST |