missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ALS2 Partial ORF (AAH29174, 221 a.a. - 320 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Especificaciones

Número de acceso AAH29174
Para utilizar con (aplicación) Antibody Production, ELISA, Protein Array, Western Blot
Formulación 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
ID de gen (Entrez) 57679
Peso molecular 36.74kDa
View More Specs

Products 2
Product Code Brand Cantidad Price Quantity & Availability  
Product Code Brand Cantidad Price Quantity & Availability  
16112893
View Documents Promotion Details
Abnova™
H00057679-Q01.25UG
25 ug
508.00€
25 microgramos
View alternative products
Save Up to 
»
Fecha estimada de envío: 10-05-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16102893
View Documents Promotion Details
Abnova™
H00057679-Q01.10UG
10 ug
335.00€
10 microgramos
View alternative products
Save Up to 
»
Fecha estimada de envío: 10-05-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Description

Description

The protein encoded by this gene contains an ATS1/RCC1-like domain, a RhoGEF domain, and a vacuolar protein sorting 9 (VPS9) domain, all of which are guanine-nucleotide exchange factors that activate members of the Ras superfamily of GTPases. The protein functions as a guanine nucleotide exchange factor for the small GTPase RAB5. The protein localizes with RAB5 on early endosomal compartments, and functions as a modulator for endosomal dynamics. Mutations in this gene result in several forms of juvenile lateral sclerosis and infantile-onset ascending spastic paralysis. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: PSQDLKPVPERCNQCSQLLITMTDKEDHVIISDSHCCPLGVTLTESQAENHASTALSPSTETLDRQEEVFENTLVANDQSVATELNAVSAQITSSDAMSS
Specifications

Specifications

AAH29174
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ALS2CR6/ALSJ/FLJ31851/IAHSP/KIAA1563/MGC87187/PLSJ
ALS2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
57679
ALS2 (Human) Recombinant Protein (Q01)
PSQDLKPVPERCNQCSQLLITMTDKEDHVIISDSHCCPLGVTLTESQAENHASTALSPSTETLDRQEEVFENTLVANDQSVATELNAVSAQITSSDAMSS
RUO
ALS2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications