Learn More
Abnova™ Human AIP Partial ORF (NP_003968.1, 156 a.a. - 249 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009049-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a receptor for aryl hydrocarbons and a ligand-activated transcription factor. The encoded protein is found in the cytoplasm as part of a multiprotein complex, but upon binding of ligand is transported to the nucleus. This protein can regulate the expression of many xenobiotic metabolizing enzymes. Also, the encoded protein can bind specifically to and inhibit the activity of hepatitis B virus. [provided by RefSeq]
Sequence: KVESPGTYQQDPWAMTDEEKAKAVPLIHQEGNRLYREGHVKEAAAKYYDAIACLKNLQMKEQPGSPEWIQLDKQITPLLLNYCQCKLVVEEYYEEspecificaciones
NP_003968.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.08kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KVESPGTYQQDPWAMTDEEKAKAVPLIHQEGNRLYREGHVKEAAAKYYDAIACLKNLQMKEQPGSPEWIQLDKQITPLLLNYCQCKLVVEEYYE | |
RUO | |
AIP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9049 | |
AIP (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARA9/FKBP16/FKBP37/SMTPHN/XAP2 | |
AIP | |
Recombinant | |
wheat germ expression system |