missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AGPAT3 Partial ORF (NP_064517, 155 a.a. - 259 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00056894-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16198836

  • 302.00€ / 10 microgramos


Fecha estimada de envío
23-10-2020
Añadir a la cesta

Descripción

Descripción

Sequence: VVEGLRRLSDYPEYMWFLLYCEGTRFTETKHRVSMEVAAAKGLPVLKYHLLPRTKGFTTAVKCLRGTVAAVYDVTLNFRGNKNPSLLGILYGKKYEADMCVRRFP
Especificaciones

Especificaciones

NP_064517
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.29kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VVEGLRRLSDYPEYMWFLLYCEGTRFTETKHRVSMEVAAAKGLPVLKYHLLPRTKGFTTAVKCLRGTVAAVYDVTLNFRGNKNPSLLGILYGKKYEADMCVRRFP
RUO
AGPAT3
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
56894
AGPAT3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LPAAT-GAMMA1|MGC4604
AGPAT3
Yes
Videos
Documentos