missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ADAMTS7 Partial ORF (NP_055087, 1589 a.a. - 1686 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00011173-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16103836

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 07-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene is a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) family. Members of this family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene contains two C-terminal TS motifs. [provided by RefSeq]

Sequence: VQRRLVKCVNTQTGLPEEDSDQCGHEAWPESSRPCGTEDCEPVEPPRCERDRLSFGFCETLRLLGRCQLPTIRTQCCRSCSPPSHGAPSRGHQRVARR
Especificaciones

Especificaciones

NP_055087
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VQRRLVKCVNTQTGLPEEDSDQCGHEAWPESSRPCGTEDCEPVEPPRCERDRLSFGFCETLRLLGRCQLPTIRTQCCRSCSPPSHGAPSRGHQRVARR
RUO
ADAMTS7
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
11173
ADAMTS7 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ADAM-TS7/DKFZp434H204
ADAMTS7
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales