missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ACTA2 Full-length ORF (AAH17554.1, 1 a.a. - 377 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00000059-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MCEEEDSTALVCDNGSGLCKAGFAGDDAPRAVFPSIVGRPRHQGVMVGMGQKDSYVGDEAQSKRGILTLKYPIEHGIITNWDDMEKIWHHSFYNELRVAPEEHPTLLTEAPLNPKANREKMTQIMFETFNVPAMYVAIQAVLSLYASGRTTGIVLDSGDGVTHNVPIYEGYALPHAIMRLDLAGRDLTDYLMKILTERGYSFVTTAEREIVRDIKEKLCYVALDFENEMATAASSSSLEKSYELPDGQVITIGNERFRCPETLFQPSFIGMESAGIHETTYNSIMKCDIDIRKDLYANNVLSGGTTMYPGIADRMQKEITALAPSTMKIKIIAPPERKYSVWIGGSILASLSTFQQMWISKQEYDEAGPSIVHRKCFEspecificaciones
AAH17554.1 | |
AP, Array, ELISA, WB-Re | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
67.21kDa | |
Glutathione Sepharose 4 Fast Flow | |
10μg | |
RUO | |
ACTA2 | |
Wheat Germ (in vitro) | |
GST Tag |
wheat germ expression system | |
Liquid | |
59 | |
Human ACTA2 Full-length ORF (AAH17554.1, 1 a.a. - 377 a.a.) Recombinant Protein with GST-tag at N-terminal | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at −80°C. Aliquot to avoid repeated freezing and thawing. | |
AAT6, ACTSA | |
ACTA2 | |
Yes |