missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AANAT Partial ORF (NP_001079.1, 1 a.a. - 79 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00000015-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16104204

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Arylalkylamine N-acetyltransferase belongs to the superfamily of acetyltransferases. It is the penultimate enzyme in melatonin synthesis and controls the night/day rhythm in melatonin production in the vertebrate pineal gland. Melatonin is essential for seasonal reproduction, modulates the function of the circadian clock in the suprachiasmatic nucleus, and influences activity and sleep. This enzyme is rapidly inactivated when animals are exposed to light at night. This protein is 80% identical to sheep and rat AA-NAT. Arylalkylamine N-acetyltransferase may contribute a multifactorial genetic diseases such as altered behavior in sleep/wake cycle. [provided by RefSeq]

Sequence: MSTQSTHPLKPEAPRLPPGIPESPSCQRRHTLPASEFRCLTPEDAVSAFEIEREAFISVLGVCPLYLDEIRHFLTLCPE
Especificaciones

Especificaciones

NP_001079.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.43kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MSTQSTHPLKPEAPRLPPGIPESPSCQRRHTLPASEFRCLTPEDAVSAFEIEREAFISVLGVCPLYLDEIRHFLTLCPE
RUO
AANAT
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
15
AANAT (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AA-NAT/SNAT
AANAT
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales