Learn More
HSPB8/HSP22 Antibody (3C12-H11), Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals NBP1-48303
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18213893
  • 511.87€ / 0.10 miligramo
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
HSPB8/HSP22 Monoclonal specifically detects HSPB8/HSP22 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
HSPB8/HSP22
Monoclonal
1 mg/mL
Western Blot 1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Alpha-crystallin C chain, CRYAC, E2IG1E2-induced gene 1 protein, H11DHMN2, heat shock 22kDa protein 8, heat shock 27kDa protein 8, heat shock protein beta-8, HMN2, HMN2A, HSP22CMT2L, HspB8, Protein kinase H11, Small stress protein-like protein HSP22
Mouse
Protein G purified
RUO
Primary
Detects approx 22kDa. Does not cross react with alpha crystallin.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1 κ
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
3C12-H11
Unconjugated
Q9UJY2
HSPB8
His-tagged human recombinant HSP22
0.1 mg
Protein Kinase
26353
Human, Mouse, Rat
Purified