Learn More
Invitrogen™ HSPA4 (HSP70) Human ELISA Kit
Sandwich ELISA Kit
Marca: Invitrogen™ EHHSPA4X10
Detalles adicionales : Peso : 2.50000kg
Descripción
The Human HSPA4 (HSP70) ELISA quantitates Hu HSPA4 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu HSPA4. Principle of the method The Human HSPA4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The HSP 70 family is composed of four highly conserved proteins: HSP 70, HSC 70, GRP 75 and GRP 78. HSP 70 expression is strongly induced in response to heat stress. HSP 70 and HSC 70 play key roles in the cytosolic endoplasmic reticulum and mitochondrial import machinery and are found in both the cytosol and nucleus of mammalian cells. GRP 78 is localized in the endoplasmic reticulum, where it receives imported secretory proteins and is involved in the folding and translocation of nascent peptide chains. GRP 75 expression is restricted to the mitochondrial matrix and aids in the translocation and folding of nascent polypeptide chains of both nuclear and mitochondrial origin. It has been postulated that members of the HSP 70 family act as force-generating motors, relying on the hydrolysis of ATP for their activity.Especificaciones
P34932 | |
2 ng/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Supernatant | |
10 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
3308 | |
<12% | |
Pre-coated 96 well plate, Standard, Stop Solution, Adhesive Plate Covers, Chromogen, Wash Buffer, SAV-HRP, Assay Diluent concentrate, Biotinylated Detection Antibody | |
10 x 96 Tests | |
Cell Cycle & Proliferation | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
2.469 to 600 ng/mL | |
2 ng/mL to 600 ng/mL | |
Biotin | |
Mitochondrial Proteins | |
This ELISA pair antibody also can recognize mouse and rat HSP70 | |
Human | |
Colorimetric Microplate Reader | |
APG-2,HEL-S-5 A,HS24/P52,HSPH2,RY,hsp70,hsp70RY | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL | |
APG-2, HEL-S-5 A, HS24/P52, HSPH2, RY, hsp70, hsp70RY | |
1 hr. 20 min. |