missing translation for 'onlineSavingsMsg'
Learn More

HSPA2 Mouse anti-Human, Mouse, Rat, Clone: 1A4A9, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66291-1-IG-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16818035

  • 109.00€ / 20 microlitros

Envío estimado: 29-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This antibody well recognized the endogenous HSPA4 protein in testis and multiple cell lines.

In cooperation with other chaperones, Hsp70s stabilize preexistent proteins against aggregation and mediate the folding of newly translated polypeptides in the cytosol as well as within organelles. These chaperones participate in all these processes through their ability to recognize nonnative conformations of other proteins. They bind extended peptide segments with a net hydrophobic character exposed by polypeptides during translation and membrane translocation, or following stress-induced damage.
Especificaciones

Especificaciones

HSPA2
1A4A9
Unconjugated
HSPA2
Liquid
HSPA2
HSPA2 Fusion Protein Ag24228
Protein G
RUO
Antibody
Monoclonal
Mouse, Human, Rat
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P14659, P17156, P54652
Heat shock 70 kDa protein 2, heat shock 70kDa protein 2, HSP70 2, HSP70 3, HSPA2
Mouse
IgG1
20 μL
-20°C
Primary
15512, 3306, 60460
Videos
SDS
Documentos
Certificados