missing translation for 'onlineSavingsMsg'
Learn More

Hsp90A/B Antibody (Hyb-K41220A), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NB110-96870AF488

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215514

  • 611.22€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Hsp90A/B Monoclonal specifically detects Hsp90A/B in Human, Mouse, Rat, Yeast samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

Hsp90A/B
Monoclonal
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
HSP90AA1
Recombinant human HSP90alpha; Specificity mapped to amino acids 291-304
0.1 mL
3320
Human, Mouse, Rat, Yeast
Purified
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Hyb-K41220A
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38
Mouse
Protein G purified
Primary
Detects 90kD proteins corresponding to the molecular mass of hsp90 alpha or beta.
Store at 4C in the dark.
IgG2a
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales