missing translation for 'onlineSavingsMsg'
Learn More

HSDL2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP2-87606

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213728

  • 455.71€ / 100 microlitros
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

HSDL2 Polyclonal specifically detects HSDL2 in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

HSDL2
Polyclonal
Unconjugated
PBS, 2% Sucrose
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
chromosome 9 open reading frame 99, EC 1.1.1, EC 1.1.1.100, FLJ25855, hydroxysteroid dehydrogenase like 2, member 1, MGC10940
The immunogen is a synthetic peptide directed towards the middle region of human HSDL2
100 μL
Lipid and Metabolism
84263
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales