missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10768-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein containing a potential clathrin-binding motif, consensus dileucine signals, and tyrosine-based sorting signals for targeting to vesicles of lysosomal lineage. The encoded protein may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 3. Alternate splice variants exist, but their full length sequence has not been determined.Especificaciones
HPS3 | |
Polyclonal | |
Unconjugated | |
HPS3 | |
Hermansky Pudlak syndrome 3, HPS3, SUTAL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12807, 310288, 84343 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q91VB4, Q969F9 | |
HPS3 | |
HPS3 Fusion Protein Ag1209 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |