missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ HPD Protein

Highly purified. Generating reliable and reproducible results.

Marca:  Novus Biologicals™ NBP1-49443-0.05MG

Detalles adicionales : Peso : 0.00970kg

 Ver más versiones de este producto

Código de producto. 18215913

  • 516.19€ / 0.05 miligramo
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-393 of Human HPD The Recombinant Human HPD Protein is derived from E. coli. The Recombinant Human HPD Protein has been validated for the following applications: SDS-Page.
Especificaciones

Especificaciones

NP_002141
ELISA, SDS-PAGE, Western Blot
3242
Protein
Recombinant human HPD protein, fused to His-tag at N-terminus, expressed in E. coli
Store at -80°C. Avoid freeze-thaw cycles.
HPD
Recombinant
1mg/mL
20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 20% glycerol, 50mM NaCl
47kDa
0.05mg
HPD, 1-393aa, Human, His tag, E. coli. (NP_002141).
RUO
Unconjugated
>90%
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales