missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDEXX Water HPC for Quanti-Tray
IDEXX Water HPC for Quanti-Tray is an accurate test for determining heterotrophic plate counts in a 100ml sample.
en la oferta
Especificaciones
Color | Rosa |
---|---|
Para utilizar con (equipo) | Quanti-Tray |
Tamaño de muestra | 100 mL |
Requisitos de almacenamiento | Almacenar entre 2 y 30 °C, lejos de la luz |
Método de prueba | Cuantificación |
Descripción
- Heterotrophic plate count made easy for bottling and manufacturing plants and on-premise testing
- Quantifies heterotrophic organisms in a 100 mL format
- No autoclaving, filtering, or agar
- No colony counting or interpretation
- Convenient 100-mL sample size—test source water, transport vehicle water, manufacturing line water, and finished products
- Simple procedure needs no special training
- Shares the same Quanti-Tray system as Colilert, Enterolert, and Pseudalert tests for maximum efficiency
- Results valid at 44–72 hours
Especificaciones
Rosa | |
100 mL | |
Cuantificación | |
Recuento de placas heterotróficas |
Quanti-Tray | |
Almacenar entre 2 y 30 °C, lejos de la luz | |
44 h |