Learn More
HOXC9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA566648
Detalles adicionales : Peso : 0.01000kg
Descripción
Immunogen sequence: DSLISHDNEDL LASRFPATGA HPAAARPSGL VPDCSDFPSC SFAPKPAVFS Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
HOXC9 Polyclonal antibody specifically detects HOXC9 in Human samples. It is validated for Immunocytochemistry.Especificaciones
HOXC9 | |
Polyclonal | |
Unconjugated | |
HOXC9 | |
homeo box C9; homeobox C9; homeobox protein Hox-3.2; Homeobox protein Hox-3B; Homeobox protein Hox-C9; HOX3; Hox-3.2; HOX3B; Hoxc8; Hoxc9; HOXC-9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3225 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P31274 | |
HOXC9 | |
Recombinant Human HOXC9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only