Learn More
Sperian Protection™ Honeywell™ Shoelace
Product Code. 13487406
Change view
Click to view available options
Length (Metric):
750mm
1500mm
Unit Size:
25 pairs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
13487406 750mm 25 pairs
13477406 1500mm 25 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13487406 Supplier Sperian Protection™ Supplier No. 6000601
  • ONLINE EXCLUSIVE PRICE
    125.24€
    69.37€ / 25 pares
    Ahorro 55.87€
    44% Descuento
Fecha estimada de envoi: 16-12-2026